Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Conductivity TDS Meter, Datalogging, Sper Scientific
Supplier: SPER SCIENTIFIC
850039
97011-778EA 674.22 USD InStock
97011-778
Conductivity TDS Meter, Datalogging, Sper Scientific
Electrochemical Meters Conductivity Meters
Records data in the plant or field for later computer storage and analysis.
Automatically records up to 16,000 data points in the field along with time and date. These can later be downloaded to a computer via the meter’s RS232 port and the results viewed instantly on software which comes with the unit. The software also enables real-time datalogging directly from the meter to the computer. Simultaneously displays conductivity or TDS and temperature. A user adjustable ATC coefficient enables absolute (non-temperature compensated) measurements, as well as accurate measurements in solutions other than water. Features manual or auto-range, min/max, calendar/clock, auto power off, and hold function. This well-made unit has a detachable probe, fold-out easel back, and a large, easy-to-read LCD.
Ordering information: Comes ready to use in a protective foam-lined carrying case, complete with probe, software, RS232 computer connection cable, 4 AA batteries, and one 3V button cell battery. Five year warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty