Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Total Trihalomethanes (THM) Reagent Set, Hach
Supplier: Hach
2790800 2790800
89231-192EA 906.68 USD InStock
CA11024-876 89231-192
Total Trihalomethanes (THM) Reagent Set, Hach
Environmental Test Kits
For determination of Total Trihalomethanes by the THM Plus™ Method.
  • Screen drinking water for regulated disinfection byproducts (DBP).
  • Fast results
  • Cost effective
  • Sensitive
Hach Method 10132. Range: 10-600 μg/L CHCl3. Reagent Set includes: THM Plus™ Reagent 1, 2, 3 Solutions and THM Plus 4 Powder Pillows for 50-99 tests. Use as a screening tool to optimize treatment processes, to locate areas of high THM levels in distribution systems and to determine levels of trihalomethanes in drinking water samples.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty