Nine-Parameter Test Kit, Model FF-1A, Hach
Supplier: Hach
243002 243002
89230-938EA 505.82 USD
89230-938 CA11024-822
Nine-Parameter Test Kit, Model FF-1A, Hach
Environmental Test Kits
Maintaining good water quality is essential for profitable fish production.
  • An economical choice, this model includes nine basic water quality tests
  • Reagents and equipment are supplied for dropcount titration and colorimetric measurement using continousgradient color discs
In many cases, the availability and cost of "crop" insurance is directly related to testing programs. This kit utilizes drop count titration and color disc methods to deliver fast, accurate results.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.