Sound Meter, Sper Scientific
840005
57800-210EA 184.67 USD
57800-210
Sound Meter, Sper Scientific
Easy to operate with analog display.
Small hand-held unit can be used to measure sound levels anywhere: measure sound levels in the workplace; quality control for sound level inspection of products; environmental noise pollution; background sound level, traffic, and more.
The two scales cover the 40-80dB and 80-120dB ranges, with an accuracy of ±3.0 dB. Simple to operate; one selection dial with battery test mode and easy-to-read results. Stable, reliable operation with signal oscillator calibration and a capacitance microphone. Based on the "A" decibel frequency weighting.
Certifications: CE approved.
Ordering information: Comes with a carrying case, instructions and a 9V battery. 5 year warranty.