Would you like to log in again?
Zinc Reagent Set, Zincon, 20 mL, Hach
Supplier: Hach
2429300 2429300
97014-598EA 91 USD InStock
97014-598 CA2429300
Zinc Reagent Set, Zincon, 20 mL, Hach
Environmental Test Kits
For determination of Zinc by the ZincoVer® Zincon method
USEPA approved for wastewater analysis (digestion is required for determining total zinc) using Hach Method 8009. Sample Size: 20 mL. Range: 0.01 - 3.00 mg/L Zn. Set contains ZincoVer 5 Powder Pillows and Cyclohexanone for 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty