ManVer® 2 Hardness Indicator Powder Pillows, Hach
Supplier: Hach
92899 92899
89230-896PK 57.06 USD
89230-896 CA928-99
ManVer® 2 Hardness Indicator Powder Pillows, Hach
Environmental Test Kits
ManVer® 2 (Calmagite) Hardness Indicator, for determination of hardness and chelants by titration.
Ordering information: Pack of 100 powder pillows.