Force Gauge with RS232 Output, Sper Scientific
Supplier: Sper Scientific
840060
14003-192EA 1117.59 USD
14003-192
Force Gauge with RS232 Output, Sper Scientific
Dynamometers
With this force gauge, you can measure tension or compression anywhere or mount it on the force gauge test stand and connect with software 850080 via the RS232 port for automated testing and process control
Display results right side up or upside down for compression or tension applications. The peak hold function also works in both modes. Also features a zero (tare) function, fast or slow response option, overload protection, and a low battery indicator. Dimensions: 9” x 3.5” x 1.5” (227 x 83 x 39 mm). Weight: 20 oz (569 g).
Ordering information: Comes in a protective carrying case containing an extension rod with a full line of adapters and 6 AAA batteries.