Total Hardness Test Kit, Model 5-EP, Hach
Supplier: Hach
145400 145400
97026-828EA 49.8 USD
97026-828 CA97026-828
Total Hardness Test Kit, Model 5-EP, Hach
Environmental Test Kits
Water hardness is caused almost entirely by calcium and magnesium ion.
  • Utilizes drop count titration a simple, economical titration method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Other di- and trivalent metals have a similar effect, but usually are not present in high enough concentration in potable waters to cause problems. Hardness increases soap consumption in laundries and causes scale in boilers. With the Model 5-EP test kit, simply count the number of drops required to change the solution color to determine hardness concentration. Utilizes drop count titration with a range of 1-30 gpg.
Ordering information: Contains approximately 100 tests.