Stopwatch and Countdown Timer, Sper Scientific
Supplier: Sper Scientific
810025R 810025RC
97035-858EA 66.15 USD
97035-858 97035-856
Stopwatch and Countdown Timer, Sper Scientific
Timers Digital Timers
The unique feature of this timer is that it also has a true stopwatch mode with ¹/₁₀₀ second resolution, lap and split with a range of 100 hours.
  • Has built-in stand, pocket clip, and magnet
In the Timer mode it counts down from 100 hours in 1 second intervals. The display indicates time elapsed after the alarm and a memory recalls last timer setting for reuse.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-858.
Ordering information: Five year warranty.
Delivery information: Button cell battery included.