Snap-A-Gels™ Tris-Tricine Mini Gels, Jule
420T10M15 420T10M10 12T10M15T 1020T10M10T 10T75M10 12T10M15 12T10M10T 16T75M10 1020T10M10 420T10M10T 16T15M2 1020T75M10 10T75M15 1020T75M15 1020T15M2 1020T10M15T 16T75M15 1020T10M15 420T10M15T 12T10M10 10T15M2
10820-572PK 25.34 USD
10820-572 10820-562 14222-478 10820-570 14222-476 14222-484 10820-566 14222-474 10820-522 14222-482 10820-520 10820-564 14222-472 14222-468 14222-480 10820-514 14222-470 10820-512 10820-568 10820-518 10820-516
Snap-A-Gels™ Tris-Tricine Mini Gels, Jule
These Tris-Tricine, polyacrylamide, precast mini gels, called Snap-A-Gels™, are available in many single and gradient concentrations
The Tris-Tricine precast mini gels can be used for denatured samples. Run denatured samples by using denaturing sample buffer and SDS in the Running Buffer. As in the Laemmli System, the Tricine is in the running buffer. Running Buffer is available from VWR International. The Tricine Gels are run with a different buffer in the upper and lower chambers of your electrophoresis apparatus. Manufacturer offers single concentration gels with a 4% stacking gel and the most precise linear gradients on the market. Manufacturer has been selling gels since 1986.
Manufacturer's special plastic material does not inhibit polymerization of acrylamide, so the cassettes behave like glass. The combination of manufacturer's special material and proprietary process for producing gels enables manufacturer to provide precast gels of the highest quality. The gold tinted cassettes make it easier to see the color blue, therefore, loading sample wells and viewing the tracking dye is easier.
Jule Snap-A-Gels are available with a variety of options to suit your sample requirementst. For example, the gels come in two thicknesses of 0.75mm (for better blotting and faster staining), and 1.50 mm (for loading more sample and greater gel strength). The Gels can be purchased with 10 wells, 15 wells, or with a flat top to accommodate an IPG strip plus one reference well.
Jule Snap-A-Gels fit the Following apparatus: Amersham-Pharmacia-Hoefer: SE250, SE260 Mighty Small; Bio-Rad Corporation: Protean II, III, 360, Tetra, Transblot & one Jule gel will fit in the III Dodeca; Invitrogen-Novex: Surelock, Xcell I and II; EX Apparatus-Thermo: EC-120; CBS Scientific: MGV-302 (one gel), MGV-402; Owl/SS/Enprotech: ISS Enprotech unit (one gel), Daiichi Devices; Owl Scientific: Penguin P8DS, Wolverine P82 (needs adapter), P81 Puffin (single sided) and Fisher Biotech: FB-VE10-1, FB-VE121 (needs adapter).
Although Tris-Tricine Gels are ideal for separating proteins as small as 2 to 3 kDa (Kilodaltons), they can be used for proteins ranging from 220 to 2 kDa. Choose Tricine Gels to get optimum separations from 70 kDa to 2 kDa. Tricine Gel buffer is 1.0 M Tris-HCl, pH 8.45, 0.1% SDS. Whether you have a simple or complex sample, the Jule Gels are priced to satisfy your budget without sacrificing performance.
Ordering information: Jule precast mini gels are sold in packs of 10 gels. A twin pack is available for purchase by contacting your VWR sales representative. The Jule Gels will fit most popular running apparatuses.