Perimatic™ Premier Pump Dispenser, Jencons
JENC256-005
53519-880EA 8458.79 USD
53519-880
Perimatic™ Premier Pump Dispenser, Jencons
The Perimatic™ Premier is a heavy-duty, microprocessor-controlled precision dispensing pump, ideally suited for rapidly and reproducibly filling a variety of containers including, vials, bottles, test tubes and petri dishes without splashing or frothing
Features include memory for storing calibration values, volumes and set-ups for various tubing sizes. There is a choice of automatic dispensing with the interval timer or manual dispening (by hand or footswitch), plus sequential slow/fast/slow dispensing.
The Premier achieves its high performance due to its precision rotor and pinch valve design. The Perimatic™ Premier is particularly suited to media dispensing as its 'stand-by' mode ensures constant re-circulation of the reservoir contents, thereby keeping the media in suspension when not actively dispensing.
Ordering information: Supplied with three sets of tubing assemblies and footswitch.