VWR® Professional Infinity Planachromatic Phase Trinocular Microscope
Supplier: VWR, part of Avantor
VWR®
1100102900491
89404-894EA 3917.18 USD
89404-894
VWR® Professional Infinity Planachromatic Phase Trinocular Microscope
Microscopes
The VWR® Professional Infinity Planachromatic Phase Trinocular Microscope is suitable for advanced lab, hospital, research, and university applications.
  • Widefield eyepieces
  • Reversed quadruple nosepiece
  • Coaxial coarse and fine focusing adjustment
This VWR® Professional microscope features a Siedentopf type trinocular head with 30° inclination and 360° rotating capabilities. Light distribution is 100/0, 20/80. Additional standard features include widefield eyepieces WF 10X/20mm, reversed quadruple nosepiece, infinity planachromatic objective PL 4X, phase contrast infinity planachromatic objectives PLPh 10X,40X S, and 100X S-Oil, coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, special 5-hole turret type Abbe condenser 1.25 N.A. with choices for phase contrast 10X, 40X, and 100X, brightfield and darkfield, Centering telescope, and Koehler illumination 3W LED with intensity control. Main power supply is 110V.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.