VWR® Stereo Zoom Boom Stand Trinocular Microscope
1100200500591
89404-486EA 5929.32 USD InStock
89404-486
VWR® Stereo Zoom Boom Stand Trinocular Microscope
Microscopes Stereo Microscopes
The VWR® Stereo Zoom Boom Stand Trinocular Microscope is a high-performance stereo microscope designed for life science, industrial and pharmaceutical inspection, electronic assembly, and bonding operations.
  • Trinocular stereo microscope head
  • Widefield eyepieces
  • Includes boom stand
The VWR® Stereo Zoom Boom Stand Trinocular Microscope features the Greenough Zoom Optical System, a 35° trinocular head with 360° rotation, photo/video: 100% [right eyetube], and widefield eyepieces 10X/F.N. 23. The unit offers 6.7:1 zoom ratio, WD = 113mm, a magnification range of 0.75X–5X, and boom stand.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.