VWR® Professional Infinity Planachromatic Binocular Microscope
1100102900461
89404-896EA 2447.41 USD InStock
89404-896
VWR® Professional Infinity Planachromatic Binocular Microscope
Microscopes Upright Microscopes
The VWR® Professional Infinity Planachromatic Binocular Microscope is suitable for advanced lab, hospital, research, and university applications.
  • Widefield eyepieces
  • Reversed quadruple nosepiece
  • Coaxial coarse and fine focusing adjustment
This VWR® Professional microscope features a Siedentopf type binocular head with 30° inclination and 360° rotating capabilities. Additional standard features include widefield eyepieces WF 10X/20mm, reversed quadruple nosepiece, infinity planachromatic objectives IPL 4x, 10x, 40x S, and 100x S-Oil, coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, centerable and focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, and Koehler illumination 3W LED with intensity control. Main power supply is 110V.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.