VWR® Stereo Zoom Plain Binocular Microscope
Supplier: VWR, part of Avantor
1100200500521
89404-480EA 422.26 USD
89404-480
VWR® Stereo Zoom Plain Binocular Microscope
The VWR® Stereo Zoom Plain Binocular Microscope is a high-performance stereo microscope designed for life science, industrial and pharmaceutical inspection, electronic assembly, and bonding operations.
  • Binocular stereo microscope head
  • Widefield eyepieces
  • 6.7:1 zoom ratio
The microscope features the Greenough Zoom Optical System, a binocular head with 360° rotation, photo/video: 100% [right eyetube], and widefield eyepieces 10X/F.N. 23. The unit offers 6.7:1 zoom ratio, WD = 113mm, and a magnification range of 0.75X–5X.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.