VWR® Professional Trinocular Microscope
Supplier: VWR, part of Avantor
VWR®
1101001300011V 1101000300782V 1101001401781 1101002400022V 1101001401101V 1101002300042V 1100102900451
89403-568EA 19.51 USD
89403-568 89403-372 89403-334 89403-474 89403-336 89403-338 89404-886
VWR® Professional Trinocular Microscope
Microscopes
The VWR® Professional Trinocular Microscope is suitable for advanced lab and hospital applications in biological, medical, and agricultural research fields.
  • Widefield eyepieces
  • Reversed quadruple nosepiece
  • Coaxial coarse and fine focusing adjustment
This VWR® Professional microscope features a Siedentopf type trinocular head with 30° inclination and 360° rotating capability. Light distribution is 100/0, 20/80. Standard features include widefield eyepieces WF 10X/20mm, reversed quadruple nosepiece, planachromatic objectives PL 4X, 10X, 40X S, and 100X S-Oil, coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, centerable and focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, and Koehler illumination 3W LED with intensity control. Main power supply is 110V.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.