Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Standard Compound Monocular Microscope

Supplier: VWR International
1101005100101 1100102900431 1101001701221 1101001701881 1101001400572V 1101001701231 1101001702551 1101001702711 1101001402104 1101001402862 1101001401541 1101001402861 1101001704591 1101001702051 1101001703072 1101001701251 1101001704121 1101001400533V
The VWR® Standard Compound Monocular Microscope is ideal for advanced educational applications and to introduce students to the world of microscopy.

  • Widefield eyepieces
  • Reversed quintuple nosepiece
  • Coaxial coarse and fine focusing adjustment

This VWR® Standard Compound microscope features a Siedentopf type monocular head with 30° inclination and 360° rotation. Also included are widefield eyepieces WF 10X/18mm, a reversed quintuple nosepiece, achromatic objectives PL 4X, 10X, 40X S, and 100X S-Oil, coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, and built-in LED illumination with intensity control. Main power supply is 110V.

Certifications: C-UL, UL Listed. CSA certified.

Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.



 

ORDER

Expand All / Collapse All

VWR® Darkfield Accessories
Description VWR Catalog Number Unit Price Quantity
VWR® Eyecups (pair) 89496-648 Each Retrieving
VWR® Eyepiece Micrometer 89496-658 Each Retrieving
VWR® Objective 100X AC 89496-636 Each Retrieving
VWR® Objective 100X SP 89496-646 Each Retrieving
VWR® Objective 10X AC 89496-628 Each Retrieving
VWR® Objective 10X SP 89496-640 Each Retrieving
VWR® Objective 20X AC 89496-630 Each Retrieving
VWR® Objective 20X AC, VWR® Objective 40X AC 89496-632 Each Retrieving
VWR® Objective 20X SP 89496-642 Each Retrieving
VWR® Objective 40X SP 89496-644 Each Retrieving
VWR® Objective 4X AC 89496-626 Each Retrieving
VWR® Objective 4X SP 89496-638 Each Retrieving
VWR® Objective 60X AC 89496-634 Each Retrieving
VWR® Widefield Eyepiece Micrometer 89496-624 Each Retrieving
VWR® Widefield Eyepiece WF10X / 18 mm 89496-622 Each Retrieving
VWR® Widefield Eyepiece WF15X/12 mm, with Pointer 89403-328 Each Retrieving
VWR® Widefield Eyepiece WF 20X/11 mm 89403-332 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

VWR® Darkfield Accessories
Description VWR Catalog Number Unit Price Quantity
VWR® Dust Cover 89403-468 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES

Expand All / Collapse All

Services
Description Details Includes Service Coverage Out-of-Service Coverage VWR Catalog Number Unit Price Quantity
VWR® Microscope Preventative Maintenance Each device has been installed and powered up before on-site service. This service include 5 Microscope PM Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA;San Francisco, CA; Las Vegas, NV; Portland, OR; Seattle, WA Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote 76006-288 Each Retrieving
VWR® Microscope Preventative Maintenance (Add On Item) Each device has been installed and powered up before on-site service. Additional One Microscope PM service to 76006-288 (This is add on item) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA;San Francisco, CA; Las Vegas, NV; Portland, OR; Seattle, WA Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote 76019-102 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty