Would you like to log in again?
VWR® Stereo Basic Halogen/Fluorescent 2X–4X Microscope
Supplier: VWR International
1100200800561
The VWR® Stereo Basic Halogen/Fluorescent 2X–4X Microscope is designed for use in electronics inspection, education, and life science applications.
  • Binocular stereo microscope head
  • Widefield eyepieces
  • Transmitted illumination
The VWR® Stereo Basic Halogen/Fluorescent 2X–4X Microscope features a 45° binocular stereo microscope head, widefield eyepieces 10X/F.N. 20, turret objective 2X–4X magnification, 12V/10W intensity-controlled halogen incident illumination, and 5W fluorescent transmitted illumination. Main power supply is 100V–240V.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.
ORDER
Expand All / Collapse All
VWR® Microscopes
Description VWR Catalog Number Unit Price Quantity
VWR® Stereo Basic Halogen/Fluorescent 2X–4X Microscope 89404-502 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty