VWR® Stereo Basic Halogen/Fluorescent 1X–3X Microscope
Supplier: VWR, part of Avantor
1100200800551
89404-500EA 882.35 USD
89404-500
VWR® Stereo Basic Halogen/Fluorescent 1X–3X Microscope
The VWR® Stereo Basic Halogen/Fluorescent 1X–3X Microscope is designed for use in electronics inspection, education, and life science applications.
  • Binocular stereo microscope head
  • Widefield eyepieces
  • Transmitted illumination
The VWR® Stereo Basic Halogen/Fluorescent 1X–3X Microscope features a 45° binocular stereo microscope head, widefield eyepieces 10X/F.N. 20, turret objective 1X–3X magnification, 12V/10W intensity-controlled halogen incident illumination, and 5W fluorescent transmitted illumination. Main power supply is 100V–240V.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.