Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Stereo Basic Halogen 1X–3X Microscope

Supplier: VWR International
The VWR® Stereo Basic Halogen 1X–3X Microscope is designed for use in electronics inspection, education, and life science applications.

  • Binocular stereo microscope head
  • Widefield eyepieces
  • Transmitted illumination

The VWR® Stereo Basic Halogen 1X–3X Microscope features a 45° binocular stereo microscope head, widefield eyepieces 10X/F.N. 20, turret objective magnification of 1X–3X, 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100V–240V.

Ordering information: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

VWR® Microscopes
Description VWR Catalog Number Unit Price Quantity
VWR® Stereo Basic Halogen 1X–3X Microscope 89404-496 Each Retrieving

VWR® Eyepieces
Description VWR Catalog Number Unit Price Quantity
VWR® Widefield 10X/F.N. 20mm 89403-378 Each Retrieving
VWR® Widefield 15X/F.N. 13mm 89403-380 Each Retrieving
VWR® Widefield 20X/F.N. 10mm 89403-382 Each Retrieving
VWR® Widefield 30X/F.N. 8mm 89403-384 Each Retrieving
VWR® Widefield 5X/F.N. 22mm 89403-376 Each Retrieving

VWR® Micrometer Eyepieces
Description VWR Catalog Number Unit Price Quantity
VWR® Graduated Linear Line and Plain Cross Hair, Point = 0.1mm/14mm (Widefield 10X/F.N. 20) 89403-386 Each Retrieving
VWR® Graduate Linear Line, Point = 0.2mm/14mm (Widefield 10X/F.N. 20) 89403-388 Each Retrieving

VWR® Polarizing, Darkfield Equipment, and Spare Bulbs
Description VWR Catalog Number Unit Price Quantity
VWR® 12V/10W Halogen Replacement Bulb for Transmitted Illumination 89403-462 Each Retrieving
VWR® 12V/15W Halogen Replacement Bulb for Incident Illumination 89403-398 Each Retrieving
VWR® 5W Fluorescent Bulb for Incident Illumination 89403-400 Each Retrieving
VWR® B301: Polarizing Equipment (1-Piece Rotary) 89403-394 Each Retrieving
VWR® Darkfield Attachment, with Iris Diaphragm 89403-396 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty