Would you like to log in again?
HQ11d Portable Starter Package with PHC201 pH Electrode, Hach
Supplier: Hach
8507530
89233-210EA 1368.82 USD InStock
89233-210
HQ11d Portable Starter Package with PHC201 pH Electrode, Hach
Electrochemical Meters pH and Multi-Parameter Meters
Designed for your water applications, the Hach HQ11d portable meter is an advanced meter that takes the guesswork out of measurements.
  • Portable meter measures critical water quality parameters without the need for multiple instruments
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements IntelliCAL™ smart probes store all calibrations in the probe
  • Designed for demanding conditions
  • Convenient kit includes everything you need to start testing
HQd™ meters connect with IntelliCAL™ probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty