Hydrogen Sulfide Color Disc Test Kit, Model HS-WR, Hach
Supplier: Hach
223801 223801
89231-270PK 413.99 USD
89231-270 CA11024-952
Hydrogen Sulfide Color Disc Test Kit, Model HS-WR, Hach
Environmental Test Kits
Resulting from the anaerobic decomposition of organic matter, hydrogen sulfide is present in many water supplies.
  • Simple and fast color disc method
  • Wide range measurement for maximum flexibility
  • Includes a Pretreatment Kit for turbid or colored samples
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Sewage and industrial wastes are other sources of sulfide pollution. Highly toxic, this compound has a characteristic rotten egg odor, which can be detected long before harmful concentrations are reached. Sulfide reacts with colorimetric reagents to form a blue color, which is measured with a color disc.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.