Sodium Periodate Powder Pillows for Manganese, 10 ml, Hach
Supplier: Hach
2107769 2107769
89231-312PK 77.13 USD
CA11016-476 89231-312
Sodium Periodate Powder Pillows for Manganese, 10 ml, Hach
Environmental Test Kits
For determination of manganese by the periodate oxidation method
EPA approved for reporting wastewater analyses (digestion required). Hach method 8034.
Ordering information: Pack of 100 Powder Pillows.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.