Nessler Reagent, 100 ml, Hach
Supplier: Hach
2119432 2119432
89231-320PK 72.35 USD
CA21194-32 89231-320
Nessler Reagent, 100 ml, Hach
Environmental Test Kits
For ammonia nitrogen determination by the Nessler method (Hach method 8038)
EPA accepted for wastewater analysis (distillation required). Range: 0.02-2.50 mg/L NH3-N. Also used in visual test kits - alternate ranges apply.
Ordering information: Contains mercury. Dispose of according to local, state and federal regulations.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.