Data Logging Sound Level Meter, Model HD600, Extech
HD600 HD600-NIST
97014-500EA 878.41 USD
97014-500 10171-854
Data Logging Sound Level Meter, Model HD600, Extech
Sound Meters
Datalogging Sound Level Meter that stores up to 20,000 records with real date and time stamp
The HD600 features datalogging capability up to 20,000 records at speed from 1/sec to 1/59sec and can capture up to 10 reading/sec when connected to a PC. Other features include MIN/MAX, Data Hold, Auto power off, Built-in USB port, and double molded housing with built-in stand. AC/DC analog output. A and C weighting.
Certifications: CE marked.
Ordering information: Complete with AC adaptor, Windows® compatible software, USB cable, wind cover, tripod, built-in stand, hard carrying case, and 9V battery. Three year warranty.