Personal Noise Dosimeter, Extech
SL355
89370-206EA 1039.33 USD
89370-206
Personal Noise Dosimeter, Extech
Compact, lightweight meter with USB interface
The SL355 can perform OSHA and IEC Noise accumulation surveys with Adjustable Criterion Level, Exchange Rate, and Threshold. UBS Interface. Includes Windows® compatible software to control setup and retrieve/analyze stored data. Applications include personal accumulated noise exposure measurement, workplace noise assessment, OSHA and other regulatory agency compliances.
Certifications: CE marked.
Ordering information: Complete with belt clip, clip-on 0.5" microphone with 2.6ft (0.8m) cable, 3 AAA batteries, mini screwdriver, software, USB cable, and carrying case. One year warranty.