Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
DPD Free Chlorine Reagent, Test 'N Tube Vials, Hach
Supplier: Hach
2105545 2105545
97026-566EA 79.16 USD InStock
CA97026-566 97026-566
DPD Free Chlorine Reagent, Test 'N Tube Vials, Hach
Environmental Test Kits
For free chlorine determination by the DPD Free Chlorine method.
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Test 'N Tube Reagent Vials contain pre-measured DPD reagent
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Range: 0.09 - 5.00 mg/L as Cl2.
Ordering information: Pack of 50 vials.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty