Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "bromine-1-mol-l-in-acetic-acid"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Avantor Performance Materials
Description:   Standardization at 25 [degree]C traceable to NIST Standard ReferenceMaterial. The actual lot standardization value, accurate to within ± 1part per 1000, is provided on the label. Normality: Actual Value Reported.