    Sort by:
  • Search Within Results
Sodium 1-tridecanesulfonate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   [Reagent for Ion-Pair Chromatography] CAS Number: 5802-89-1 MDL Number: MFCD00024993 Molecular Formula: C13H28O3S Molecular Weight: 286.41 Purity/...