    Sort by:
Ethyl propyl sulphide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (TCE0224-010G)
Supplier:  TCI America
Description:   CAS Number: 4110-50-3 MDL Number: MFCD00039940 Molecular Formula: C5H12S Molecular Weight: 104.21 Purity/Analysis Method: 96.0% (GC) Form: Cl...