Would you like to log in again?
ACE® Excel C18 Ultra-Inert, 2 µm, Analytical UHPLC Columns, USP - L1, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
EXL1010203U EXL1013503U EXL1010546U EXL1017503U EXL1011203U EXL1013502U EXL1011003U EXL1011502U EXL1017502U EXL1011046U EXL1011002U EXL1010346U EXL1010302U EXL1010303U EXL1011546U EXL1013546U EXL1010502U EXL1010202U EXL1011246U EXL1010246U EXL1017546U EXL1011503U EXL1010503U EXL1011202U
101671-744EA 930.29 USD InStock
101671-744 101671-756 101671-764 101671-768 101671-732 101671-754 101671-726 101671-736 101671-766 101671-728 101671-724 101671-752 101671-748 101671-750 101671-740 101671-758 101671-760 101671-742 101671-734 101671-746 101671-770 101671-738 101671-762 101671-730
ACE® Excel C18 Ultra-Inert, 2 µm, Analytical UHPLC Columns, USP - L1, Mac Mod
Chromatography Columns
ACE® Excel C18 2 micron UHPLC columns use a proprietary bonding and end-capping process to produce a very rugged and robust octadecyl (C18) phase
The ultra-inert characteristics of the ACE® Excel C18 Columns make them the ideal choice for separating polar, moderately polar, and non-polar analytes (including uncharged acids and bases, strong acids or bases in un-ionized form or in ionized form using an ion-pair modifier.) ACE® Excel columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds.
ACE® Excel C18 is available in 2, 3 and 5 micron particles sizes. ACE® Excel C18 has a surface area of 300 m2/g and a carbon load of 15.5%. ACE® Excel C18 has a recommended pH range of 2-8 (working outside these ranges will reduce column lifetime).
ACE® Excel UHPLC columns are extremely robust even when operated at high pressures and temperatures. ACE® Excel 2 micron columns are packed at extreme pressures in specialized hardware. They can be operated at pressures as high as 1,000 bar. A specially engineered 2 micron inlet frit makes them less prone to plugging than conventional UHPLC columns. ACE® Excel columns are compatible with all HPLC, UPLC and UHPLC instruments.
Ordering information: ACE® Excel 2 micron UHPLC columns currently do not have a guard column system. Manufacturer highly recommends UltraShield pre-column filters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty