ACE® Excel Phenyl Ultra-Inert, 3 and 5 µm, Analytical UHPLC Compatible Columns, USP - L11, Mac Mod
EXL1150503U EXL1257503U EXL1151203U EXL1251503U EXL1157502U EXL1253546U EXL1250303U EXL1153503U EXL1251246U EXL1152502U EXL1250246U EXL1253502U EXL1151546U EXL1250346U EXL1250503U EXL1152546U EXL1251502U EXL1251203U EXL1151503U EXL1251002U EXL1150202U EXL1150546U EXL1252502U EXL1251202U EXL1151202U EXL1150246U EXL1253046U EXL1250302U EXL1151246U EXL1153546U EXL1250203U EXL1257546U EXL1253503U EXL1257502U EXL1250502U EXL1251003U EXL1250202U EXL1150502U EXL1150203U EXL1253002U EXL1151502U EXL1150346U EXL1252503U EXL1253003U EXL1251546U EXL1157503U EXL1151003U EXL1157546U EXL1250546U EXL1151046U EXL1151002U EXL1251046U EXL1152503U EXL1150303U EXL1150302U EXL1252546U EXL1153502U
101718-958EA 818.66 USD
101718-958 101718-518 101718-956 101718-516 101718-510 101718-994 101718-992 101718-514 101718-954 101718-998 101718-512 101718-996 101719-008 101718-990 101718-550 101719-000 101719-002 101719-004 101719-006 101718-506 101718-988 101718-548 101718-504 101719-010 101718-508 101718-542 101718-982 101718-980 101718-540 101718-986 101718-546 101718-502 101718-984 101718-544 101718-500 101719-012 101718-538 101718-978 101718-532 101718-972 101718-498 101718-530 101718-970 101718-976 101718-536 101718-534 101718-974 101718-968 101718-528 101718-526 101718-966 101718-960 101718-520 101718-524 101718-964 101718-962 101718-522
ACE® Excel Phenyl Ultra-Inert, 3 and 5 µm, Analytical UHPLC Compatible Columns, USP - L11, Mac Mod
ACE® Excel Phenyl 3 micron and 5 micron UHPLC compatible columns use a proprietary bonding and end-capping process to produce a very rugged and robust phenylethyl phase
ACE® Excel Phenyl has a hydrophobicity that is between C4 and C8 phases, with increased polar selectivity. ACE® Excel Phenyl has improved performance, stability and reproducibility compared to conventional phenyl phases. ACE® Excel Phenyl is compatible with highly aqueous mobile phases.
ACE® Excel Phenyl is available in 2, 3 and 5 micron particles sizes. ACE® Excel Phenyl has a surface area of 300 m2/g and a carbon load of 9.5%. ACE® Excel Phenyl has a recommended pH range of 2-8 (working outside these ranges will reduce column lifetime).
ACE® Excel UHPLC columns are extremely robust even when operated at high pressures and temperatures. ACE® Excel 3 micron and 5 micron columns are packed at extreme pressures in specialized hardware. They can be operated at pressures as high as 1,000 bar. A specially engineered 2 micron inlet frit makes them less prone to plugging than conventional UHPLC columns. ACE® Excel columns are compatible with all HPLC, UPLC and UHPLC instruments.
Ordering information: If a guard column is required for ACE® Excel 3 micron and 5 micron UHPLC compatible columns, manufacturer recommends stand-alone guard holder, and the corresponding, appropriately sized ACE® guard cartridge.