AluVer® 3 Aluminum Reagent Powder Pillows, Hach
Supplier: Hach
1429099 1429099
89230-962PK 135.69 USD
89230-962 CA1429099
AluVer® 3 Aluminum Reagent Powder Pillows, Hach
Environmental Test Kits
For aluminum determination by the aluminon method
Hach Method 8012. Range: up to 0.80 mg/L Al.
Ordering information: Pack of 100 powder pillows.