Molybdenum Reagent Set, HR, Hach
Supplier: Hach
2243400 2243400
89231-342PK 188.17 USD InStock
89231-342 CA11014-440
Molybdenum Reagent Set, HR, Hach
Environmental Test Kits
For determination of Molybdenum by the Mercaptoacetic Acid Method (Hach method 8036)
Range: Up to 35.0 mg/L as Mo6+.
Ordering information: Set Contains: MolyVer® 1, 2 and 3 Molybdenum Reagent Powder Pillows for 100 tests.