Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Hardness 1 Buffer Solution, 500 mL, Hach
Supplier: Hach
424-49 42449
97026-664EA 37.22 USD InStock
CA424-49 97026-664
Hardness 1 Buffer Solution, 500 mL, Hach
Environmental Test Kits
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C
Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty