Would you like to log in again?
ACE® Excel C8 Ultra-Inert, 2 µm, Analytical UHPLC Columns, USP - L7, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
EXL1020302U EXL1023546U EXL1020246U EXL1020203U EXL1023503U EXL1020546U EXL1020503U EXL1021246U EXL1021546U EXL1020303U EXL1021502U EXL1021046U EXL1021203U EXL1027546U EXL1021003U EXL1023502U EXL1020346U EXL1021202U EXL1027503U EXL1021002U EXL1021503U EXL1020502U EXL1027502U
101717-910EA 982.38 USD InStock
101717-910 101717-944 101717-908 101717-906 101717-942 101717-920 101717-918 101717-932 101717-938 101717-912 101717-934 101717-926 101717-930 101717-950 101717-924 101717-940 101717-914 101717-928 101717-948 101717-922 101717-936 101717-916 101717-946
ACE® Excel C8 Ultra-Inert, 2 µm, Analytical UHPLC Columns, USP - L7, Mac Mod
Chromatography Columns
ACE® Excel C8 2 micron UHPLC columns use a proprietary bonding and end-capping process to produce a very rugged and robust octyl (C8) phase
The ultra-inert characteristics of the ACE® Excel C8 Columns make them the ideal choice for separating polar, moderately polar, and non-polar analytes (including uncharged acids and bases, strong acids or bases in un-ionized form or in ionized form using an ion-pair modifier.) ACE® Excel columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds.
ACE® Excel C8 is available in 2, 3 and 5 micron particles sizes. ACE® Excel C8 has a surface area of 300 m2/g and a carbon load of 9.0%. ACE® Excel C8 has a recommended pH range of 2-8 (working outside these ranges will reduce column lifetime).
ACE® Excel UHPLC columns are extremely robust even when operated at high pressures and temperatures. ACE® Excel 2 micron columns are packed at extreme pressures in specialized hardware. They can be operated at pressures as high as 1,000 bar. A specially engineered 2 micron inlet frit makes them less prone to plugging than conventional UHPLC columns. ACE® Excel columns are compatible with all HPLC, UPLC and UHPLC instruments.
Ordering information: ACE® Excel 2 micron UHPLC columns currently do not have a guard column system. Manufacturer highly recommends UltraShield pre-column filters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty