Would you like to log in again?
BR250 Video Borescope/Wireless Inspection Camera, Extech
Supplier: Extech
BR250
97049-980EA 516.45 USD InStock
97049-980
BR250 Video Borescope/Wireless Inspection Camera, Extech
Cameras Borescopes
Video Borescope/Wireless Inspection Camera with a 9mm camera diameter and a 3.5" Color TFT LCD Wireless Monitor
The BR250 features a mini water-proof (IP67) camera head for high resolution viewing with two bright LED lamps with dimmer to illuminate viewed object. The 2GB micro SD memory card (included) captures images (JPEG) or video (AVI) for viewing on your PC. Other Features include a glare-free close-up field of view and a detachable display for easy storage.
Certifications: CE, FCC
Ordering information: Complete with 4AA batteries, rechargeable display battery, micro SD card with adaptor, USB and video cable, extension tools, 100-240V AC adaptor, stand and case. 1 yr warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty