BR200 Video Borescope/Wireless Inspection Camera, Extech
BR200
97049-978EA 435.08 USD
97049-978
BR200 Video Borescope/Wireless Inspection Camera, Extech
Video Borescope/Wireless Inspection Camera with a 17mm camera diameter and a 3.5" Color TFT LCD Wireless Monitor
The BR200 features a mini water-proof (IP67) camera head for high resolution viewing with two bright LED lamps with dimmer to illuminate viewed object. The 2GB micro SD memory card (included) captures images (JPEG) or video (AVI) for viewing on your PC. Other features include a glare-free close-up field of view and a detachable display for easy storage.
Certifications: CE, FCC
Ordering information: Complete with 4 AA batteries, micro SD memory card, SD adaptor, USB cable, extension tools, video cable, AC adaptor, magnetic base stand and storage case. 1 yr warranty.