HDV640W HD VideoScope with Wireless Handset/Articulating Probe, Extech
Supplier: Extech
HDV640W
89200-732EA 5625.32 USD
89200-732
HDV640W HD VideoScope with Wireless Handset/Articulating Probe, Extech
Cameras
Inspection Camera with Wireless Handset & 6mm Semi-Rigid, Articulating (240°) Probe (1m)
The HDV640W features a 5.7" color LCD TFT with high definition 640 x 480 VGA pixel resolution. The wireless transmitter allows you to transmit video up to 100ft (30m) from the measurement point to your monitor. It has a compact, high-resolution water-proof camera probe with a camera head that has four built-in bright LED lamps with dimmer to illuminate viewed object. The SD memory card included can store more than 15,000 images. Other features include video recording (up to 4 hours) with voice annotation and an AV output for viewing images and video directly on a monitor.
Certifications: CE, IP67
Ordering information: Includes wireless handset with articulating probe, SD card, 3.7V rechargable Li-Polymer battery, patch cable, 100/240V AC adaptor, USB and AV cables, and carrying case. 1 yr warranty.