FLIR E60 Compact Infrared Thermal Imaging Camera, FLIR
FLIR E60
89200-764EA 10483.05 USD InStock
89200-764
FLIR E60 Compact Infrared Thermal Imaging Camera, FLIR
Cameras Thermal Imaging Cameras
Compact Infrared Thermal Imaging Camera with a 320 x 240 IR Resolution that measures temperature up to 1202°F (650°C)
The FLIR E60 features a built-in laser pointer, 3.5" Touch-Screen LCD display, and a 3.1 Megapixel visible light digital camera with bright LED lamp for poorly lit environments. It has multiple measurement modes up to 3 movable Spots, up to 3 Areas (Min/Max/Average), Auto Hot/Cold spot marker, Isotherm (Above/Below), Delta T. Other features include Thumbnail Image Gallery, METERLiNK™ Bluetooth® Wireless communication, Video Recording/Streaming and Memory card slot for image storage.
Certifications: CE, IP54
Ordering information: Complete with Memory card, Li-Ion rechargeable battery, power supply, USB and Video cable, hand strap, lens cap and hard carrying case. Includes FLIR Tools software for analysis of radiometric images.
2/5/10 warranty (Camera-2 years, Battery-5 years, Detector- 10 years).