Inspection Camera with 5.5mm Flexible, 60° FOV, Macro Probe (1m)
The HDV610 features a 5.7" color LCD TFT with high definition 640 x 480 VGA pixel resolution. It has a compact, high-resolution water-proof camera probe with a camera head that has four built-in bright LED lamps with dimmer to illuminate viewed object. The SD memory card included can store more than 15,000 images. Other features include video recording (up to 4 hours) with voice annotation and an AV output for viewing images and video directly on a monitor.
Certifications: CE, IP67
Ordering information: Includes handset & camera probe, SD card, 3.7V rechargable Li-Polymer battery, patch cable, 100/240V AC adaptor, USB and AV cables, and carrying case. 1 yr warranty.