Would you like to log in again?
CalVer® 2 Calcium Indicator Powder Pillows, 50 mL sample, Hach
Supplier: Hach
85299 852-99
Indicator for determination of Calcium by EDTA titration
Powder pillows for 50-mL samples.
Ordering information: Pack of 100 powder pillows.
ORDER
Expand All / Collapse All
Description Range Size Chemical Method No. of Tests Supplier No. VWR Catalog Number Unit Price Quantity
CalVer® 2 Calcium Indicator Powder Pillows, 50 mL sample, pk/100 10 - 4000 mg/l CaCO₃ 50 mL Sample EDTA 100 85299 89230-888 Pack of 100 Retrieving Restricted
Product available on GSA Advantage®
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Method EDTA Titration
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty