Generic Pocket Colorimeter II Instruments are intended for applications not included in standard analysis systems.
- Single Parameter go anywhere portable photometer
- USEPA approved for drinking water and wastewater
- FactoryProgrammed
- Waterproof, Lightweight, Rugged
- Backlit LCD Display, Data Logging, Recall
They are available in a range of wavelengths appropriate for many custom calibrations. In addition to the factory absorbance calibration (0-2.5 A), the instruments offer two user-entered, two-point, linear calibrations, and provide a choice of either 0.00 to 2.55 or 0.0 to 99.0 for the display value.
Ordering information: 2 years warranty.