EC Medium MPN Tubes, Hach
Supplier: Hach
1410415 1410415
97026-340PK 82.34 USD
97026-340 CA14104-15
EC Medium MPN Tubes, Hach
Media for Microbiology Prepared Media for Microbiology
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.
  • Detect coliform
  • MPN method
  • Save time and money
Save time and labor costs with ready to use MPN tubes, with minimal training requirements.
Ordering information: 15/pk.