Would you like to log in again?
Chlorine, Hardness, Iron, and pH Test Kit, Model CN-39WR, Hach
Supplier: Hach
223002
89231-266PK 303.57 USD InStock
89231-266
Chlorine, Hardness, Iron, and pH Test Kit, Model CN-39WR, Hach
Environmental Test Kits
Chlorine, Hardness, Iron, and pH Test Kit combines the most popular water treatment test into one compact kit.
  • Includes frequently measured water quality parameters in one rugged test kit
  • Uses simple, economical drop count titration and color disc methods
The chlorine test is USEPA-accepted for drinking water testing if approved by individual states.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty