Would you like to log in again?
Total Organic Carbon (TOC) Reagent Set, HR, Hach
Supplier: Hach
2760445 2760445
97026-422PK 649.86 USD InStock
CA97026-422 97026-422
Total Organic Carbon (TOC) Reagent Set, HR, Hach
Environmental Test Kits
For determination of Total Organic Carbon (TOC) by the High Range Test 'N Tube™ method.
  • Affordable
  • Easy to use
  • Accurate
Hach Method 10128. Range:100 - 700 mg/L C. Set includes digestion vials and reagents for 25-50 tests (dependent upon the number of reagent blank tests performed). Requires digestion.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty