ZincoVer® 5 Zinc Reagent Powder Pillows, Hach
Supplier: Hach
21066-69 1403268 2106669
10765-196PK 23.9 USD
CA21066-69 10765-196 97026-642
ZincoVer® 5 Zinc Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of zinc by the ZincoVer® 5 zincon method
USEPA approved for wastewater analysis - Hach method 8009. Range: 0.01 to 3.00 mg/L Zn.