PhosVer® 3 Phosphate Reagent Powder Pillows, 25 mL sample, Hach
Supplier: Hach
212528 2125-28
97026-636PK 468.98 USD InStock
97026-636 CA2125-28
PhosVer® 3 Phosphate Reagent Powder Pillows, 25 mL sample, Hach
Environmental Test Kits
For phosphate (phosphorus) determination by the PhosVer 3 ascorbic acid method
Hach method 8048.
Ordering information: Pack of 1000 powder pillows.