Molybdate 3 Reagent, 100mL, Hach
Supplier: Hach
199532 1995-32
89231-256PK 30.48 USD
89231-256 CA1995-32
Molybdate 3 Reagent, 100mL, Hach
Environmental Test Kits
For low and ultra-low range silica determination by the Heteropoly Blue method
Hach methods 8282 and 8186.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.