Ward's® Immune Response: Antigen/Antibody Reactions Kit
Supplier: Avantor
WARD470190-620 WARD470015-326
470190-620KT 71.89 USD
470190-620 470015-326
Ward's® Immune Response: Antigen/Antibody Reactions Kit
Educational Classroom Kits and Activities
Who can donate blood to you?
  • Ward's Simulated Blood Is Safe & Nontoxic
  • Activities: Determining Case Histories & Safety for Blood Donations
  • Kit Utilizes Structures & Properties (LS1) & Heredity (LS3)
  • Material for 10 Setups
  • Assessments & Vocabulary Words Target Common Core Standards
Students determine which of four possible donors could safely donate blood to a patient. They also develop a case history of a mother and her children to determine the safety of her pregnancy. In the process, students employ the framework standards of Structures and Properties (LS1) and Heredity (LS3) to identify antigen and antibody reactions, how those reactions affect the body's immune response, and the basics of ABO and Rh blood typing.
This kit is aligned with all published National Standards. Pre and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core standards. The teacher's guide contains specific "Did You Know" concepts that link the real world with the scientific standards explored. The kit contains a student copymaster and material for 10 setups.
Ordering information: This kit includes Simulated Blood Samples One through Four, simulated Anti-A serum, simulated Anti-B serum, simulated Anti-Rh serum, blood-typing slides, toothpicks, a teacher's guide, and a student copymaster.